Order Entry
Italy
ContactUsLinkComponent
(S)-4-(4-Methoxybenzyloxy)-1,3-butanediol ≥96.0% (by GC)
(S)-4-(4-Methoxybenzyloxy)-1,3-butanediol ≥96.0% (by GC)
Codice catalogo TCIAM1790-500MG
Supplier:  TCI
CAS Number:  
(S)-4-(4-Methoxybenzyloxy)-1,3-butanediol ≥96.0% (by GC)
Codice catalogo TCIAM1790-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 96.0 %
  • Specific rotation [a]20/D:
    -5.0 to -11.0 deg (C=1, Methanol)

Specification Test Results

About this item

 :
 :226.27 g/mol
CAS Number:1820579-68-7
Formula:C₁₂H₁₈O₄