Order Entry
Italy
ContactUsLinkComponent
 
Neu5GcAc[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥97.0% (by HPLC)
  TCIAM1763-200MG
 :  TCI
 :  
undefined
Neu5GcAc[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥97.0% (by HPLC)
  TCIAM1763-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

 

 

 :
 :1116.09 g/mol
 :Freezer
 :C₅₆H₆₁NO₂₃