Order Entry
Italy
ContactUsLinkComponent
Neu5Ac[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥95.0% (by HPLC)
Neu5Ac[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥95.0% (by HPLC)
Catalog # TCIAM1761-200MG
Supplier:  TCI
CAS Number:  
undefined
Neu5Ac[1Me,4789Ac]α(2-6)Gal[24Bz,3Bn]-β-MP ≥95.0% (by HPLC)
Catalog # TCIAM1761-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
 :1058.05 g/mol
Storage Temperature:Refrigerator
Formula:C₅₄H₅₉NO₂₁