Order Entry
Italy
ContactUsLinkComponent
Neu5Troc[1Me,4789Ac]ɑ(2-3)Gal[26Bn]-β-MP ≥98.0% (by HPLC)
Neu5Troc[1Me,4789Ac]ɑ(2-3)Gal[26Bn]-β-MP ≥98.0% (by HPLC)
Catalog # TCIAM1729-1G
Supplier:  TCI
CAS Number:  
Neu5Troc[1Me,4789Ac]ɑ(2-3)Gal[26Bn]-β-MP ≥98.0% (by HPLC)
Catalog # TCIAM1729-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

About this item

 :
 :1073.31 g/mol
Storage Temperature:Refrigerator
CAS Number:610763-72-9
Formula:C₄₈H₅₆Cl₃NO₂₀