
2-Methyl-4,5-diphenylthiazole ≥97.0% (by GC)
Catalog # TCIAM1705-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:50.0 to 54.0 deg-C
Specification Test Results
About this item
:
:251.35 g/mol
EINECS:223-161-0
:GHS07
CAS Number:3755-83-7
Formula:C₁₆H₁₃NS
MDL Number:MFCD00186396