Order Entry
Italy
ContactUsLinkComponent
Phenylacetaldehyde for synthesis, Sigma-Aldrich®
Phenylacetaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Phenylacetaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    32 - 34 °C
  • Molar Mass:
    120.15 g/mol
  • Boiling Point:
    192 - 195 °C (1013 hPa)
  • Vapor Pressure:
    0.35 hPa (19 °C)
  • Flash Point:
    87 °C
  • Density:
    1.026 - 1.030 g/cm3 (20 °C)
  • Hill Formula:
    C8H8O

Specification Test Results

Related Information

Frequently Bought Together