4-Methoxyphenyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥95.0% (by HPLC)
Catalog # TCIAM1615-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Specific rotation [a]20/D:+59.0 to +63.0 deg (C=1,CHCl3)
Specification Test Results
About this item
:
:595.65 g/mol
Storage Temperature:Freezer
CAS Number:129575-89-9
Formula:C₃₅H₃₃NO₈