Order Entry
Italy
ContactUsLinkComponent
Methyl-4-(1,1-dioxothiomorpholino)benzoate ≥98.0% (by GC)
Methyl-4-(1,1-dioxothiomorpholino)benzoate ≥98.0% (by GC)
Catalog # TCIAM1587-1G
Supplier:  TCI
CAS Number:  
Methyl-4-(1,1-dioxothiomorpholino)benzoate ≥98.0% (by GC)
Catalog # TCIAM1587-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Solubility in hot Methanol:
    very faint turbidity

Specification Test Results

About this item

 :
 :269.32 g/mol
CAS Number:451485-76-0
Formula:C₁₂H₁₅NO₄S