
Methyl-3-(chlorosulphonyl)thiophene-2-carboxylate ≥90.0% (by GC, titration analysis)
Catalog # TCIAM1404-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 90.0 %
- Purity (Argentmetric Titration):min. 90.0 %
Specification Test Results
About this item
:
Melting Pt:63°C
UN:3261
ADR:8,III
:240.6845 g/mol
:GHS05
CAS Number:59337-92-7
Formula:C₆H₅ClO₄S₂
MDL Number:MFCD00068160