Methyl-β-cyclodextrin (mixture of several Methylated)
Supplier: TCI
Certificates
About this item
CAS Number:128446-36-6
:
MDL Number:MFCD00074980
:1303.3000 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Specific rotation [a]20/D:+150.0 to +165.5 deg (C=1.5, H2O) (calcd.on dried substance)
- Solubility in Water:almost transparency
Specification Test Results
Related Information
Frequently Bought Together