


Mesityl oxide ≥85.0% (by GC) (contains 4-Methyl-4-penten-2-one)
Supplier: TCI
Certificates
About this item
CAS Number:141-79-7
:
Formula:C₆H₁₀O
MDL Number:MFCD00008900
Merck Index:14,05908
Flash Pt:25°C
EINECS:205-502-5
UN:1229
ADR:3,III
:98.14 g/mol
:GHS02
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 85.0 %
Specification Test Results
Frequently Bought Together