

Bis(4-nitrobenzyl)malonate ≥98.0% (by HPLC, titration analysis)
TCIAM1181-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ester Value):min. 98.0 %
- Melting point:87.0 to 91.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:374.31 g/mol
:GHS07
:GHS08
:67245-85-6
:C₁₇H₁₄N₂O₈
:MFCD00191671