Order Entry
Italy
ContactUsLinkComponent
4,5-Methylenedithio-1,3-dithiol-2-one ≥95.0% (by HPLC)
4,5-Methylenedithio-1,3-dithiol-2-one ≥95.0% (by HPLC)
Codice catalogo TCIAM1154-100MG
Supplier:  TCI
CAS Number:  
4,5-Methylenedithio-1,3-dithiol-2-one ≥95.0% (by HPLC)
Codice catalogo TCIAM1154-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

About this item

 :
CAS Number:85720-62-3