Order Entry
Italy
ContactUsLinkComponent
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0% (by GC, titration analysis) for determination of the optical purity of alcohols and amines
Corrosive
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0% (by GC, titration analysis) for determination of the optical purity of alcohols and amines
Supplier:  TCI
Certificates
Corrosive
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99.0% (by GC, titration analysis) for determination of the optical purity of alcohols and amines
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Optical purity (GC):
    min. 99.0 ee%
  • Functionality test:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together