Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3,3-Dimethyloctane ≥98%

Flammable
Irritant
3,3-Dimethyloctane ≥98%
Certificates
3,3-Dimethyloctane ≥98%
3,3-Dimethyloctane ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 4110-44-5 Molecular Formula: C10H22 Formula Weight: 142.29 Boiling Point: 161° UN No.: 3295 MDL No.: MFCD00048874

  • CAS Number:4110-44-5
  • Formula:C₁₀H₂₂
  • MDL Number:MFCD00048874
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:161 °C
  • Melting Pt:–69 °C
  • Density:0.74
  • Storage Temperature:Ambient
  • UN:3295
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 37,90 €
    37,90 €
    1 * 1 g
       37,90 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 138,00 €
    138,00 €
    1 * 5 g
       138,00 €