Order Entry
Italy
ContactUsLinkComponent
3-Ethoxy-N,N-diethylaniline ≥97.0% (by GC, titration analysis)
3-Ethoxy-N,N-diethylaniline ≥97.0% (by GC, titration analysis)
  TCIAM1039-25G
 :  TCI
 :  
undefined
3-Ethoxy-N,N-diethylaniline ≥97.0% (by GC, titration analysis)
  TCIAM1039-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

 

 

 :
 :193.29 g/mol
 :270°C
 :217-469-4
 :GHS07
 :1864-92-2
 :C₁₂H₁₉NO
 :MFCD00043936