Order Entry
Italy
ContactUsLinkComponent
N-(2-Methoxyethyl)methylamine ≥97.0% (by GC, titration analysis)
Flammable
Corrosive
N-(2-Methoxyethyl)methylamine ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
N-(2-Methoxyethyl)methylamine ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together