D-Mannono-1,4-lactone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:26301-79-1
:
Formula:C₆H₁₀O₆
MDL Number:MFCD00065020
:178.14 g/mol
EINECS:247-596-0
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:151.0 to 156.0 deg-C
- Specific rotation [a]20/D:+49.0 to +54.0 deg (C=2, H2O)
- Solubility in Water:within almost transparency
Specification Test Results
Frequently Bought Together