

2-Methyl-2-butenenitrile ≥70% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4403-61-6
:
Formula:C₅H₇N
MDL Number:MFCD00037169
:81.12 g/mol
Boiling Pt:50°C
Flash Pt:–17°C
EINECS:224-544-5
UN:3273
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 70.0 %
Specification Test Results
Frequently Bought Together