
4-(Methylthio)-m-cresol ≥98.0% (by GC)
Catalog # TCIAM0531-25G
Supplier: TCI
CAS Number:
Specifications
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Brown to Dark purple powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:53.0 to 57.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:154.23 g/mol
Formula:C₈H₁₀OS
MDL Number:MFCD00045773