Order Entry
Italy
ContactUsLinkComponent
(-)-Terpinen-4-ol ≥95.0% (by GC) (contains 30% (+)-Terpinen-4-ol at maximum)
Irritant
(-)-Terpinen-4-ol ≥95.0% (by GC) (contains 30% (+)-Terpinen-4-ol at maximum)
Catalog # TCIAM0319-25ML
Supplier:  TCI
CAS Number:  
Irritant
(-)-Terpinen-4-ol ≥95.0% (by GC) (contains 30% (+)-Terpinen-4-ol at maximum)
Catalog # TCIAM0319-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %
  • Enantiomeric ratio (GC):
    (-) >70, (+) <30

Specification Test Results

About this item

 :
 :154.25 g/mol
Boiling Pt:210°C
 :GHS07
CAS Number:20126-76-5
Formula:C₁₀H₁₈O
MDL Number:MFCD00167108