2,2'-Methylenebis(6-tert-butyl-p-cresol) ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2,2'-Methylenebis(4-methyl-6-tert-butylphenol), 2,2-Methylenebis(6-tert-butyl-p-cresol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane, 2,2'-Methylenebis(6-tert-butyl-4-methylphenol), 3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethane
CAS Number:119-47-1
:
Formula:C₂₃H₃₂O₂
MDL Number:MFCD00043641
:340.51 g/mol
EINECS:204-327-1
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:130.0 to 134.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together