Lomitapide ≥98.0% (by HPLC)
: TCI
:182431-12-5
:
:C₃₉H₃₇F₆N₃O₂
:MFCD16620494
:693.73 g/mol
:142°C
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):5.75 to 6.35 %
Frequently Bought Together