



Lithium-tert-butanolate ∼10% ∼1 mol/L in THF
Catalog # TCIAL0253-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Concentration (Neutralization titration):9.6 to 10.2 w/w%
Specification Test Results
About this item
:
:80.06 g/mol
Density:0.89
EINECS:217-611-5
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:1907-33-1
Formula:C₄H₉LiO
MDL Number:MFCD00050479