Calcium levulinate dihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAL0043-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 % (calcd.on anh.substance)
- Water:10.0 to 13.0 %
- Melting point:121.0 to 125.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:270.29 g/mol
Merck Index:14,01679
EINECS:209-725-9
CAS Number:5743-49-7
Formula:C₅H₈O₃
MDL Number:MFCD00045959