Order Entry
Italy
ContactUsLinkComponent
N,N'-[(2S,2'S)-[(2-Iodo-1,3-phenylene)bis(oxy)]bis(propane-2,1-diyl)]bis(mesitylamide) ≥96.0% (by HPLC)
N,N'-[(2S,2'S)-[(2-Iodo-1,3-phenylene)bis(oxy)]bis(propane-2,1-diyl)]bis(mesitylamide) ≥96.0% (by HPLC)
  TCIAI1122-200MG
 :  TCI
 :  
N,N'-[(2S,2'S)-[(2-Iodo-1,3-phenylene)bis(oxy)]bis(propane-2,1-diyl)]bis(mesitylamide) ≥96.0% (by HPLC)
  TCIAI1122-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Optical purity (LC):
    min. 98.0 ee%
  • Melting point:
    212.0 to 216.0 deg-C

 

 

 :
 :642.58 g/mol
 :1399008-27-5
 :C₃₂H₃₉IN₂O₄