Isoindolin-1-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:480-91-1
:
Formula:C₈H₇NO
MDL Number:MFCD03085939
:133.15 g/mol
Boiling Pt:337°C
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:149.0 to 153.0 deg-C
Specification Test Results
Frequently Bought Together