Order Entry
Italy
ContactUsLinkComponent
 
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)
 :  TCI
 
(S,S)-2,2'-Isopropylidenebis(4-isopropyl-2-oxazoline) ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Optical purity (LC):
    min. 98.0 ee%

 

Frequently Bought Together