Order Entry
Italy
ContactUsLinkComponent
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane ≥92.0% (by GC)
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane ≥92.0% (by GC)
Catalog # TCIAI1044-25G
Supplier:  TCI
CAS Number:  
undefined
2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane ≥92.0% (by GC)
Catalog # TCIAI1044-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 92.0 %

Specification Test Results

About this item

 :
 :784.12 g/mol
Boiling Pt:194°C
CAS Number:26738-51-2
Formula:C₁₄HF₂₉O₄
MDL Number:MFCD00054717