
IR-775 chloride ≥90.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:199444-11-6
:
Formula:C₃₂H₃₆Cl₂N₂
MDL Number:MFCD03427067
:519.55 g/mol
:GHS07
Specification Test Results
- Appearance:Green to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 90.0 %
- Lambda max.:772.0 to 777.0nm (MeOH)
- Molar extinction coefficient:min. 240000 (MeOH, 772.0 to 777.0 nm)
Specification Test Results
Frequently Bought Together