
5-Iodo-1-methyl-1H-pyrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34091-51-5
:
Formula:C₄H₅IN₂
MDL Number:MFCD11109327
:208.00 g/mol
:GHS07
Specification Test Results
- Purity (GC):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
Specification Test Results
Frequently Bought Together