Order Entry
Italy
ContactUsLinkComponent
2-Methylpropene ∼10% in isopropyl ether
Flammable
Irritant
Health
2-Methylpropene ∼10% in isopropyl ether
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
2-Methylpropene ∼10% in isopropyl ether
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (HNMR):
    10.0 to 12.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together