
Imidazo[1,5-a]pyridine-3-carboxaldehyde ≥98.0% (by titrimetric analysis)
TCIAI0754-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Brown powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
:
:146.15 g/mol
:GHS07
:56671-66-0
:C₈H₆N₂O
:MFCD08752619