2-Isobutyl-4,5-dimethyl-3-thiazoline (mixture of isomers) ≥95.0% (by GC)
Catalog # TCIAI0737-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
Specification Test Results
About this item
:
:171.30 g/mol
Boiling Pt:70°C
Flash Pt:87°C
EINECS:265-969-6
CAS Number:65894-83-9
Formula:C₉H₁₇NS
MDL Number:MFCD01680822