

1,1'-Isopropylidenezirconocene dichloride ≥96.0% (by titrimetric analysis)
Catalog # TCIAI0645-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Chelometric Titration):min. 96.0 %
Specification Test Results
About this item
:
:332.38 g/mol
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:138533-79-6
Formula:C₁₃H₁₄Cl₂Zr