

(R)-(+)-Indoline-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
TCIAI0589-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+114 to +117 deg (C=1, 2mol/L HCl)
:
:163.18 g/mol
:GHS07
:GHS08
:98167-06-7
:C₉H₉NO₂
:MFCD00792496