Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
 
Isopropylmagnesium bromide 15% ∼1 mol/L in THF
  TCIAI0518-250G
 :  TCI
 :  
undefined
 
 
 
 
Isopropylmagnesium bromide 15% ∼1 mol/L in THF
  TCIAI0518-250G
 :  TCI
 :  

 

  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Concentration (sec Butanol method):
    15.0 to 16.5 w/w%

 

 

 :
 :147.30 g/mol
 :213-056-8
 :3399
 :4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :920-39-8
 :C₃H₇BrMg
 :MFCD00672110