



Isobutylmagnesium bromide 17% ∼1 mol/L in THF
Catalog # TCIAI0517-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):17.0 to 18.7 %
Specification Test Results
About this item
:
:161.33 g/mol
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:926-62-5
Formula:C₄H₉BrMg
MDL Number:MFCD00075126