Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium-2-ethylhexanoate

Lithium-2-ethylhexanoate

Lithium-2-ethylhexanoate
Certificates
Lithium-2-ethylhexanoate
Lithium-2-ethylhexanoate

2 Options Available Below

Product Details & Documents

CAS No.: 15590-62-2 Molecular Formula: C8H15LiO2 Formula Weight: 150.15 Physical Form: Lump MDL No.: MFCD00058702

Synonyms:  Lithium 2-ethylhexanoate, 2-Ethylhexanoic acid lithium salt, 2-Ethylcaproic acid lithium salt
  • CAS Number:15590-62-2
  • Formula:C₈H₁₅LiO₂
  • MDL Number:MFCD00058702
  • Molecular Weight:150.15 g/mol
  • Storage Temperature:Ambient
  • EINECS:239-657-5
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.