Order Entry
Italy
ContactUsLinkComponent
Isopropyl methyl sulphide ≥97.0% (by GC)
Flammable
Isopropyl methyl sulphide ≥97.0% (by GC)
Catalog # TCIAI0483-25G
Supplier:  TCI
CAS Number:  
Flammable
Isopropyl methyl sulphide ≥97.0% (by GC)
Catalog # TCIAI0483-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :90.18 g/mol
Boiling Pt:85°C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
 :GHS02
CAS Number:1551-21-9
Formula:C₄H₁₀S
MDL Number:MFCD00015035