Order Entry
Italy
ContactUsLinkComponent
 
 
Diallyl isophthalate ≥98.0% (by GC, titration analysis)
 :  TCI
 
 
Diallyl isophthalate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Bromination Method):
    min. 98.0 %

 

 

Frequently Bought Together