Order Entry
Italy
ContactUsLinkComponent
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate >60.0% (by GC) (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate >60.0% (by GC) (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)
Supplier:  TCI
Certificates
undefined
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate >60.0% (by GC) (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 60.0 %

Specification Test Results

Frequently Bought Together