Order Entry
Italy
ContactUsLinkComponent
1,2-O-Isopropylidene-ɑ-D-glucofuranose ≥97.0% (by GC)
1,2-O-Isopropylidene-ɑ-D-glucofuranose ≥97.0% (by GC)
Catalog # TCIAI0400-25G
Supplier:  TCI
CAS Number:  
1,2-O-Isopropylidene-ɑ-D-glucofuranose ≥97.0% (by GC)
Catalog # TCIAI0400-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    156.0 to 162.0 deg-C
  • Specific rotation [a]20/D:
    -11.0 to -13.0 deg (C=1, H2O)

Specification Test Results

About this item

 :
 :220.22 g/mol
EINECS:242-420-9
CAS Number:18549-40-1
Formula:C₉H₁₆O₆
MDL Number:MFCD00063244