
4,4'-Isopropylidenediphenol dimethacrylate ≥98.0% (by GC)
TCIAI0389-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:73.0 to 76.0 deg-C
- Solubility in hot Methanol:almost transparency
:
:364.44 g/mol
:221-846-9
:GHS07
:3253-39-2
:C₂₃H₂₄O₄
:MFCD00008585