
Isoguanine ≥98.0% (by HPLC)
TCIAI0370-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
:
:151.13 g/mol
:222-157-6
:GHS07
:3373-53-3
:C₅H₅N₅O
:MFCD00142931