
4,4'-Isopropylidenediphenoxyacetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3539-42-2
:
Formula:C₁₉H₂₀O₆
MDL Number:MFCD00060114
:344.36 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:177.0 to 180.0 deg-C
Specification Test Results
Frequently Bought Together