

Isopropenyloxytrimethylsilane ≥90.0% (by GC) trimethylsilylating agent
Catalog # TCIAI0324-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 90.0 %
Specification Test Results
About this item
:
:130.26 g/mol
Boiling Pt:94°C
Flash Pt:-7°C
Flash Pt:–7°C
Storage Temperature:Refrigerator
EINECS:217-393-1
UN:1993
ADR:3,II
:GHS02
:GHS07
CAS Number:1833-53-0
Formula:C₆H₁₄OSi
MDL Number:MFCD00042845