
Isophthalic dihydrazide ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2760-98-7
:
Formula:C₈H₁₀N₄O₂
MDL Number:MFCD00025117
:194.19 g/mol
Melting Pt:227°C
EINECS:220-425-7
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Bromination Method):min. 95.0 %
- Solubility in hot Water:within almost transparency
Specification Test Results
Frequently Bought Together