2-(1H-Imidazol-5-yl)acetic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
: TCI
:3251-69-2
:
:C₅H₇ClN₂O₂
:MFCD00012698
:162.58 g/mol
:221-840-6
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together