Order Entry
Italy
ContactUsLinkComponent
 
 
3,3'-Iminodipropionitrile ≥98.0% (by GC)
 :  TCI
 
undefined
 
3,3'-Iminodipropionitrile ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together